missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Tissue Section Adhesive

Product Code. 15239938 Shop All Epredia Products
Change view
Click to view available options
Capacity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
15239938 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15239938 Supplier Epredia™ Supplier No. 86014

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Increase the affinity of the tissue for the slide with Epredia™ Tissue Section Adhesive: Pre-mixed and ready-to-use.

Use in the water bath to increase the affinity of the tissue for the slide.
  • Creates no background residue
  • Improves adhesion for techniques utilizing heat
TRUSTED_SUSTAINABILITY

Specifications

Capacity 500 mL
Product Type Tissue Section Adhesive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.