Learn More
Riley™ Tilla Safety Glasses
Product Code. 31702886
Change view
Click to view available options
Lens Tint:
Clear
Gray
Unit Size:
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Tint Unit Size
31702886 Clear Pack of 20
31702889 Gray Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31702886 Supplier Riley™ Supplier No. RLY00461
  • ONLINE EXCLUSIVE PRICE
    403.43 €
    136.42 € / Pack of 20
    Save 267.01 €
    66% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Over-glasses safety eyewear offering full coverage with brow and side protection, designed for comfort and compatibility with prescription glasses.
  • Can be worn over spectacles
  • Ventilated temple tips and side shields
  • Brow protection
  • Anti-fog lens
  • UV400 protection
  • Ideal to be worn by site visitors
Specifications
Certifications/Compliance CE or - UKCA or - EN 166:2001
Lens Coating Moisture Repellent, UV-treated
Lens Tint Clear
Product Type Overglasses
For Use With (Application) Component and Product Assembly, Woodwork and Fabrication, Glass and Glazing, Surveying
Frame Color Clear
Transmittance Value 92%
Weight (Metric) 44 g
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate (High-impact)
Frame Material Polycarbonate
UV Protection UV400
Frame Type Indirect Vented