missing translation for 'onlineSavingsMsg'
Learn More

Rilmazafone Hydrochloride, TRC

Product Code. 30005288
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
5 mg
Unit Size:
0.5mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005288 0.5 mg 0.5mg
30006558 5 mg 5mg
30008899 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005288 Supplier Toronto Research Chemicals Supplier No. TRCR5097150.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85815-37-8
Molecular Formula C21H20Cl2N6O3 . HCl
Molecular Weight (g/mol) 511.8
Synonym 5-[[(Aminoacetyl)amino]methyl]-1-[4-chloro-2-(2-chlorobenzoyl)phenyl]-N,N-dimethyl-1H-1,2,4-triazole-3-carboxamide Monohydrochloride, 450191S
IUPAC Name 5-[[(2-aminoacetyl)amino]methyl]-1-[4-chloro-2-(2-chlorobenzoyl)phenyl]-N,N-dimethyl-1,2,4-triazole-3-carboxamide;hydrochloride
SMILES Cl.CN(C)C(=O)c1nc(CNC(=O)CN)n(n1)c2ccc(Cl)cc2C(=O)c3ccccc3Cl

Specifications

Quantity 0.5 mg
InChI Formula InChI=1S/C21H20Cl2N6O3.ClH/c1-28(2)21(32)20-26-17(11-25-18(30)10-24)29(27-20)16-8-7-12(22)9-14(16)19(31)13-5-3-4-6-15(13)23;/h3-9H,10-11,24H2,1-2H3,(H,25,30);1H
Formula Weight 510.0741
Chemical Name or Material Rilmazafone Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.