missing translation for 'onlineSavingsMsg'
Learn More

Riluzole, TRC

Product Code. 30138070
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138070 50 mg 50mg
30133956 100 mg 100mg
30008192 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138070 Supplier Toronto Research Chemicals Supplier No. TRCR51000050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1744-22-5
Molecular Formula C8 H5 F3 N2 O S
Molecular Weight (g/mol) 234.2
Synonym 6-(Trifluoromethoxy)-2-benzothiazolamine, Benzothiazole, 2-amino-6-(trifluoromethoxy)- (7CI,8CI), 2-Amino-6-(trifluoromethoxy)benzothiazole, 6-(Trifluoromethoxy)-1,3-benzothiazol-2-amine, 6-(Trifluoromethoxy)-1,3-benzothiazol-2-ylamine, 6-(Trifluoromethoxy)-2-aminobenzothiazole, 6-Trifluoromethoxybenzo[d]thiazol-2-amine, 6-Trifluoromethoxybenzothiazol-2-ylamine, PK 26124, RP 54274, Rilutek, Rilutor, Riluzole
IUPAC Name 6-(trifluoromethoxy)-1,3-benzothiazol-2-amine
SMILES Nc1nc2ccc(OC(F)(F)F)cc2s1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C8H5F3N2OS/c9-8(10,11)14-4-1-2-5-6(3-4)15-7(12)13-5/h1-3H,(H2,12,13)
Formula Weight 234.0075
Chemical Name or Material Riluzole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.