missing translation for 'onlineSavingsMsg'
Learn More

Ansell Ringers R075 PVC-Coated Chemical Resistant/Waterproof Impact Gloves - Extended Cuff

Product Code. 16619073
Click to view available options
Size:
Small
Large
2X-Large
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 16619073

Brand: Ansell 07510

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Developed with Ringers' F3 Technology™ and patented TPR design for increased dexterity and comfort. Textured PVC coating on the palm enhances grip capabilities, while also providing chemical resistance.

  • TPR impact protection on top of the hand and over the full length of the fingers
  • PVC coating is waterproof and provides resistance to chemicals
  • Inner liner provides cut resistance throughout the entire glove
  • Durable textured palm coating enhances grip capabilities
  • Short gauntlet-style cuff keeps debris away from the hands
  • Level 1 Contact Heat performance rating at an outside exposure of 100°C, in accordance with test method EN 407
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 5
Certifications/Compliance (Europe) EN 374_2003 Microorganism, EN 420:2003 + A1:2009
Applications Industrial
Coating Material PVC
Color Hi-vis Red
Cut Resistance Rating ANSI A3
Exterior Finish Fully Textured
Latex Free No
Material DuPont™ Aramid Fibers
Product Type Chemical-resistant Gloves
Size Large
EN388 Standard 4441CP
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES KLMOPT
EN ISO 374 - 5 Yes
Glove Cuff Style Gauntlet
Series R-075
Length (English) 18 in.
Length (Metric) 456 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.