missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ RIPA 56

Product Code. 18746666
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18746666 50 mg 50mg
18708426 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18746666 Supplier R&D Systems™ Supplier No. 7339/50
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and metabolically stable RIP1 kinase inhibitor

RIPA 56 is a potent and metabolically stable receptor-interacting protein kinase 1 (RIPK1) inhibitor (EC50 = 13 nM). RIPA-56 shows no inhibition of RIP3 kinase at 10 μM . RIPA 56 blocks necroptosis and aging of the mouse male reproductive system. RIPA 56 reduces tumor necrosis factor α (TNFα)-induced mortality and multi-organ damage in a mouse model of systemic inflammatory response syndrome (SIRS). RIPA 56 is orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Hydroxy-2,2-dimethyl-N-(phenylmethyl)butanamide
CAS 1956370-21-0
Quantity 50 mg
Product Line Tocris
Target RIP Kinase Inhibitors
Molecular Formula C13H19NO2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.