missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ RIPGBM
GREENER_CHOICE

Product Code. 18767946
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18767946 10 mg 10mg
18776156 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18767946 Supplier Tocris Bioscience™ Supplier No. 6892/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

RIPK2 modulator; pro-drug of cRIPGM; orally bioavailable and brain penetrant

RIPGBM is a RIPK2 modulator, pro-drug of cRIPGM. cRIPGM binds RIPK2 (apparent Kd = 2.3 μM). Selectively induces apoptosis in glioblastoma multiforme cancer stem cell lines (EC50 is <500 nM). Five-fold selective for CSCs compared with control cell types. Reduces the formation of a prosurvival RIPK2/ TAK1 complex and increases the formation of a proapoptotic RIPK2/caspase 1 complex. Inhibits tumor formation and growth in vivo. Orally bioavailable and brain penetrant.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[1,4-Dihydro-1,4-dioxo-3-[(phenylmethyl)amino]-2-naphthalenyl]-N-[(4-fluorophenyl)methyl]acetamide
CAS 355406-76-7
Quantity 10 mg
Target RIP Kinase Inhibitors
Molecular Formula C26H21FN2O3
Formula Weight 1818
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.