missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Risdiplam
GREENER_CHOICE

Product Code. 18775955
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18775955

Brand: Tocris Bioscience™ 7916/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective SMN2 gene splicing modifier

Risdiplam is a selective survival of motor neuron-2 (SMN2) pre-mRNA splicing modifier. It potently enhances SMN2 splicing in vitro (EC1.5X = 4 nM) and increases SMN protein in both brain (up to 100%) and muscle tissues (up to 49%) of transgenic mouse models of spinal muscular atrophy (SMA). Risdiplam dose-dependently increases the number of vesicular glutamate transporter 1 (vGlut1) proprioceptive inputs on motor neurons, the number of L3 to L5 motor neurons, the percentage of fully innervated neuromuscular junctions, and muscle size in the extensor digitorum longus muscles in SMN Δ7 mice.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-(4,7-Diazaspiro[2.5 ]oct-7-yl)-2-(2,8-dimethylimidazo[1,2-b]pyridazin-6-yl)-4 H-pyrido[1,2-a]pyrimidin-4-one
CAS 1825352-65-5
Quantity 50 mg
Target DNA, RNA and Protein Synthesis Compounds
Molecular Formula C22H23N7O
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.