missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Risdiplam

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Risdiplam is a selective survival of motor neuron-2 (SMN2) pre-mRNA splicing modifier. It potently enhances SMN2 splicing in vitro (EC1.5X = 4 nM) and increases SMN protein in both brain (up to 100%) and muscle tissues (up to 49%) of transgenic mouse models of spinal muscular atrophy (SMA). Risdiplam dose-dependently increases the number of vesicular glutamate transporter 1 (vGlut1) proprioceptive inputs on motor neurons, the number of L3 to L5 motor neurons, the percentage of fully innervated neuromuscular junctions, and muscle size in the extensor digitorum longus muscles in SMN Δ7 mice.
Specifications
Specifications
| Chemical Name or Material | 7-(4,7-Diazaspiro[2.5 ]oct-7-yl)-2-(2,8-dimethylimidazo[1,2-b]pyridazin-6-yl)-4 H-pyrido[1,2-a]pyrimidin-4-one |
| CAS | 1825352-65-5 |
| Quantity | 50 mg |
| Target | DNA, RNA and Protein Synthesis Compounds |
| Molecular Formula | C22H23N7O |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido