missing translation for 'onlineSavingsMsg'
Learn More

Risedronic Acid Sodium Salt, TRC

Product Code. 30138468
Change view
Click to view available options
Quantity:
50 mg
250 mg
2.5 g
Unit Size:
2.5g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138468 250 mg 250mg
30132612 2.5 g 2.5g
30005752 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138468 Supplier Toronto Research Chemicals Supplier No. TRCR521500250MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115436-72-1
Molecular Formula C7 H10 N O7 P2 . Na
Molecular Weight (g/mol) 305.09
Synonym Phosphonic acid, P,P'-[1-hydroxy-2-(3-pyridinyl)ethylidene]bis-, sodium salt (1:1), Phosphonic acid, [1-hydroxy-2-(3-pyridinyl)ethylidene]bis-, monosodium salt (9CI), 3-Pyridyl-1-hydroxyethylidene-1,1-biphosphonic acid sodium salt, Actonel, Fossical, Gemfos, NE 58095, Risedronate sodium, Risedronic acid monosodium salt, Sodium risedronate
IUPAC Name sodium;hydroxy-(1-hydroxy-1-phosphono-2-pyridin-3-ylethyl)phosphinate
SMILES [Na+].OC(Cc1cccnc1)(P(=O)(O)O)P(=O)(O)[O-]

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C7H11NO7P2.Na/c9-7(16(10,11)12,17(13,14)15)4-6-2-1-3-8-5-6;/h1-3,5,9H,4H2,(H2,10,11,12)(H2,13,14,15);/q;+1/p-1
Formula Weight 304.983
Chemical Name or Material Risedronate sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.