missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Risperidone

Product Code. 18782594
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18782594 50 mg 50mg
18742614 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18782594 Supplier R&D Systems™ Supplier No. 2865/50
  • ONLINE EXCLUSIVE PRICE
    811.33 €
    731.85 € / 50mg
    Save 79.48 €
    9% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. 5-HT2A antagonist

Risperidone is an atypical antipsychotic agent that displays 5-HT2A receptor antagonism. Also displays high affinity at D2 receptors (Ki values are 0.4 and 3.13 nM for 5-HT2A and D2 receptors respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106266-06-2
Molecular Formula C23H27FN4O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 410.48
Purity >99%
Molecular Weight (g/mol) 410.493
Chemical Name or Material Risperidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.