missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ RITA

Product Code. 18084842
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18084842 1 mg 1mg
18023962 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18084842 Supplier R&D Systems™ Supplier No. 2443/1
  • ONLINE EXCLUSIVE PRICE
    127.00 €
    119.70 € / 1mg
    Save 7.30 €
    5% Off
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. MDM2-p53 interaction inhibitor

Anti-tumor agent that binds wild-type p53 (Kd = 1.5 nM) preventing p53-MDM2 (HDM2) interaction. Induces p53 accumulation and stimulates apoptosis in tumor cell lines expressing wild-type p53 in vitro and in vivo. Inhibits HPV-E6-mediated proteasomal degradation. Suppresses expression of cervical carcinoma HeLa xenografts in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 213261-59-7
Molecular Formula C14H12O3S2

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 292.37
Purity >97%
Molecular Weight (g/mol) 292.367
Chemical Name or Material RITA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.