missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ RKI 1447 dihydrochloride
Description
RKI 1447 dihydrochloride is a potent and selective ROCK inhibitor (IC50 values are 6.2 and 14.5 nM for ROCK2 and ROCK1 respectively); suppresses phosphorylation of ROCK substrates MLC-2 and MYPT-1 in human cancer cells, but has no effect on the phosphorylation levels of Akt, MEK, and S6 kinase. Displays anti-invasive and antitumor activities in breast cancer cells.
Specifications
Specifications
| CAS | 1782109-09-4 |
| Quantity | 50 mg |
| Product Line | Tocris |
| Product Type | RKI 1447 dihydrochloride |
| Purity | >99% |
| Molecular Weight (g/mol) | 399.29 |
| Inhibitors | RKI 1447 dihydrochloride |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?