Learn More
Invitrogen™ RNA Control 250 with Manual
The Invitrogen RNA Control 250 provides a precise, accurate, and reproducible RNA concentration reference for use in conjunction with NanoDrop™ Spectrophotometers.
211.00 € - 236.00 €
Specifications
Content And Storage | RNA Control and THE RNA Storage Solution should be stored at –20°C |
---|---|
Format | Tube |
Label or Dye | Unconjugated |
Product Line | Ambion™ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10400725
|
Invitrogen™
AM7155 |
20 μL |
211.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10713067
|
Invitrogen™
AM7155M |
2 x 20 μL, with Manual |
236.00 €
Each |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Invitrogen RNA Control 250 provides a precise, accurate, and reproducible RNA concentration reference for use in conjunction with NanoDrop™ Spectrophotometers. It is provided in two tubes containing 20 μL each. RNA Control 250 consists of a single, ultra-pure, 2000nt RNA transcript of a precisely defined sequence, molecular weight, and concentration. It is packaged in special non-stick tubes and includes the Invitrogen THE RNA Storage Solution. The RNA Control 250 will maintain ± 5% of stated concentration and >85% RNA integrity through 5 freeze/thaw cycles (10 duplicate measurements).
• A stable, highly pure transcript at a precisely known concentration
• Used to verify the accuracy of NanoDrop Spectrophotometer measurements
• Provided with THE RNA Storage Solution for blank measurements
• Enough for 20 measurements
Specifications
RNA Control and THE RNA Storage Solution should be stored at –20°C | |
Tube | |
Unconjugated | |
Ambion™ |
For Research Use Only. Not for use in diagnostic procedures.