missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Protein Solving Buffer Set PSB/TCEP
Specifications
Specifications
| Chemical Name or Material | Protein Solving Buffer |
| Quantity | 1 Set |
Safety and Handling
Signal Word
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- MIXTURE LIST-Contain: tris-(2-carboxyethyl)phosphine hydrochloride, TCEP(
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?