missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNAqueous™ Total RNA Isolation Kit

Product Code. 10075994
Change view
Click to view available options
Quantity:
50 Catridges
50 Preps
Product Type:
RNA Isolation Kit
Spin Columns
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Type unitSize
10075994 50 Preps RNA Isolation Kit Each
10176864 50 Catridges Spin Columns Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10075994 Supplier Invitrogen™ Supplier No. AM1912

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The RNAqueous Total RNA Isolation Kit is for phenol-free total RNA isolation from 1–75 mg of tissue or from 10^2–10^7 cells, using a guanidinium-based lysis/denaturant and glass fiber filter separation technology.

The RNAqueous Total RNA Isolation Kit is for phenol-free total RNA isolation from 1–75 mg of tissue or from 102–107 cells, using a guanidinium-based lysis/denaturant and glass fiber filter separation technology.

The RNAqueous kit provides:

  • Convenient syringe-based filtration
  • Compatibility with vacuum manifolds when processing multiple samples

How it works

The RNAqueous kit uses a powerful chaotropic solution for cell lysis, followed by RNA binding to glass fiber housed in a luer-lock syringe filter. Washing and elution solutions are simply pushed through the filter using a standard syringe (not included). Vacuum filtration is not required; however, the filter units are compatible with standard vacuum manifolds for processing several samples at once.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 6 mL Formaldehyde Load Dye; store at -20°C
• 60 mL Lysis/Binding Solution; 4°C
• 60 mL Water; 4°C
• 4 mL Lithium Chloride Precipitation Solution; 4°C
• 40 mL Wash Solution #1; 4°C
• 80 mL Wash Solution #2/3 (concentrated); 4°C
• 50 Filter Cartridges; room temperature
• 100 Collection Tubes; room temperature
• 10 mL Elution Solution; room temperature
Isolation Technology Spin Column (Glass Fiber Filter)
Elution Volume 50–120 μL
Sample Type Bacteria, Cells, Plant, Tissue, Yeast, Viruses, Enzymatic Reactions
Final Product Type Total RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Product Type RNA Isolation Kit
Quantity 50 Preps
Shipping Condition Room Temperature
Starting Material Amount Bacteria: ≤109 cells, Cells: ≤107, Tissue: ≤75 mg, Plant: ≤75 mg, Yeast: ≤108 cells, Virus: varies, Enzymatic Reactions: varies
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • RNAqueous Total RNA Isolation Kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
  • H371-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Lithium chloride, Formamide, Formaldehyde, Methanol, 2-Mercaptoethanol

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.