missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RNase AWAY™ Surface Decontaminant

Product Code. 10180601 Shop All MBP Molecular BioProducts Products
Change view
Click to view available options
Volume (Metric):
250 mL
475 mL
1 L
4 L
Container Type:
Bottle
Spray Bottle
Unit Size:
1 L
250mL
4 L
475mL
5 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Container Type Unit Size
10180601 1 L Bottle 1 L
11580095 250 mL Bottle 250mL
10666421 475 mL Spray Bottle 475mL
30286571 1 L Bottle 5 L
11952385 4 L Bottle 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10180601 Supplier Thermo Scientific™ Supplier No. 7003
  • ONLINE EXCLUSIVE PRICE
    105.67 €
    68.40 € / 1 L
    Save 37.27 €
    35% Off
View all promotions
 Restricted Item
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eliminate RNase and DNA from laboratory surfaces. This product reduces dependency on carcinogenic DEPC treatments and saves time spent baking glassware.

Eliminate RNase and DNA from laboratory surfaces. Thermo Scientific™ RNase AWAY™ is ideal for decontaminating apparatus, bench-tops, glassware and plasticware. It reduces dependency on carcinogenic DEPC treatments and saves time needed to bake glassware.

  • Use on pipettes, gel boxes, or RNA or DNA prep areas
  • Leaves no residue to interfere with gel polymerization or staining
  • Chemically stable and nonabrasive
  • Contains no strong acids
TRUSTED_SUSTAINABILITY

Specifications

pH 11
Composition Sodium Hydroxide with Water
Container Type Bottle
Volume (English) 33.875 oz.
Volume (Metric) 1 L
compliance-icons
Product Identifier
  • RNAse away
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.