missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ RNase-free Water

Product Code. 15879077
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15879077 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15879077 Supplier Macherey-Nagel™ Supplier No. 740378.1000
  • ONLINE EXCLUSIVE PRICE
    52.60 €
    46.62 € / 1000mL
    Save 5.98 €
    11% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

RNase-free Water to be used with NucleoSpin RNA or miRNA kits

An RNase-free working environment is a critical factor for performing successful RNA isolation and handling. Therefore, general recommendations to avoid RNase contamination should be followed:

  • Maintain a separate area, dedicated pipettors, and materials when working with RNA
  • Wear gloves when handling RNA and reagents to avoid contact with skin, which is a source of RNases
  • Change gloves frequently
  • Use sterile RNase-free plastic tubes
  • Keep all kit components sealed when not in use and all tubes tightly closed when possible

Specifications

pH 7
CAS 7732-18-5
Color Colorless
CAS Min % 0.9
Melting Point 0°C
Boiling Point 100°C
CAS Max % 1
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name water
Molecular Weight (g/mol) 18.02
Odor Odorless
Quantity 1000 mL
Specific Gravity 1.0 g/cm3
Physical Form Liquid
Chemical Name or Material Water
Grade RNase free
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.