missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ RNaseP Verification Plate, Fast 96-well

Product Code. 15880403 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 x 96 well plate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15880403 1 x 96 well plate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15880403 Supplier Applied Biosystems™ Supplier No. 4432656
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used to verify your real-time PCR instrument with Fast 96-well Block.

Confirm the performance of your instrument by running the RNase P assay. Please see your instrument user guide for full instructions on use. The Fast 96-well RNase P Verification Plate is preloaded with the necessary reagents to detect and quantitate genomic copies of the human RNase P gene, a single-copy gene encoding the RNA moiety of the RNase P enzyme. Each well contains preloaded reaction mix (1X TaqMan™ Universal PCR Master Mix, RNase P primers, and FAM™ dye-labeled probe) and template. The verification plate must demonstrate the ability to distinguish between 5,000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well. To ensure accurate results, it is recommended that the performance of your instrument be verified every six months.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at –20°C ± 5°C.

Calibrated Dye Proprietary Mix
For Use With (Equipment) ViiA™ 7 Dx System, QuantStudio™ Dx
Reaction Speed Fast
Product Type qPCR System Verification Plate
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Dry Ice
Quantity 1 x 96 well plate
Calibration Type Instrument Verification Plate
PCR Method qPCR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.