missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals RNase Removal Reagent

Product Code. 15434159
Change view
Click to view available options
Quantity:
475 mL
Unit Size:
475mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15434159 475 mL 475mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15434159 Supplier Thermo Scientific Chemicals Supplier No. J61574.R!
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

Destroys RNases and DNases on contactRNase removal reagent is used to remove the RNase from glasswares, plastic surfaces, equipments, gel boxes, benchtops and labware. It is also used to eliminate the RNase from microcentrifuge tubes without any side enzymatic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Density 1.00
For Use With (Application) Destroys RNases and DNases on contact
Product Type RNase Removal Reagent
Quantity 475 mL
Form Liquid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.