missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ro 25-6981 maleate

Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
Description
Ro 25-6981 maleate is a potent and selective activity-dependent blocker of NMDA receptors containing the GluN2B (formally NR2B) subunit. IC50 values are 0.009 and 52 μM for cloned receptor subunit combinations GluN1C/GluN2B and GluN1C/GluN2A respectively. Displays neuroprotectant effects in vivo and in vitro.
Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
Specifications
Specifications
| Chemical Name or Material | (αR,βS)-α-(4-Hydroxyphenyl)-β-methyl-4-(phenylmethyl)-1-piperidinepropanol maleate |
| CAS | 1312991-76-6 |
| Assay Percent Range | >98% |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO |
| Molecular Formula | C22H29NO2.C4H4O4 |
| Formula Weight | Observed MW: 455.55 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction