missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ro 48-8071 fumarate
GREENER_CHOICE

Product Code. 18797548
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18797548

Brand: Tocris Bioscience™ 5389/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2,3-Oxidosqualene cyclase (OSC) inhibitor; blocks cholesterol synthesis

Ro 48-8071 fumarate is a 2,3-Oxidosqualene cyclase (OSC) inhibitor (IC50 = 6.5 nM); blocks cholesterol synthesis in HepG2 cells. Reduces viability of breast cancer cell lines. Also inhibits Ebola virus (EBOV) cell entry (IC50 = 1.74 μM).
TRUSTED_SUSTAINABILITY

Specifications

CAS 189197-69-1
Quantity 10 mg
Molecular Formula (4-Bromophenyl)[2-fluoro-4-[[6-(methyl-2-propen-1-ylamino)hexyl]oxy]phenyl]methanone
Product Type Ro 48-8071 fumarate
Purity 99%
Molecular Weight (g/mol) M.W. (Observed): 564.44
Inhibitors Ro 48-8071 fumarate

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.