missing translation for 'onlineSavingsMsg'
Learn More

Robenidine Hydrochloride, TRC

Product Code. 30134607
Change view
Click to view available options
Quantity:
100 mg
10 g
100 g
Unit Size:
100g
100mg
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134607 10 g 10g
30134330 100 g 100g
17945025 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134607 Supplier Toronto Research Chemicals Supplier No. TRCR63900010G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25875-50-7
Molecular Formula C15 H13 Cl2 N5 . Cl H
Molecular Weight (g/mol) 370.66
Synonym 2,2'-Bis[(4-chlorophenyl)methylene]carbonimidic Dihydrazide Hydrochloride (1:1), Carbonimidic dihydrazide, bis[(4-chlorophenyl)methylene]-, monohydrochloride (9CI), Guanidine, 1,3-bis[(p-chlorobenzylidene)amino]-, monohydrochloride (8CI), 1,3-Bis[(p-chlorobenzylidene)amino]guanidine monohydrochloride, Cycostat, NCL 812, Robenidine hydrochloride, Robenzidene
IUPAC Name 1,3-bis[(E)-(4-chlorophenyl)methylideneamino]guanidine;hydrochloride
SMILES Cl.Clc1ccc(\C=N\NC(=N)N\N=C\c2ccc(Cl)cc2)cc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C15H13Cl2N5.ClH/c16-13-5-1-11(2-6-13)9-19-21-15(18)22-20-10-12-3-7-14(17)8-4-12;/h1-10H,(H3,18,21,22);1H/b19-9+,20-10+;
Formula Weight 369.0315
Chemical Name or Material Robenidine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.