Learn More
STEMCELL Technologies RoboSep™ HLA Chimerism Whole Blood CD19 Positive Selection Kit
Product Code. 17147781
Encompass
Click to view available options
Quantity:
60 mL Buffy Coat or Whole Blood
Unit Size:
1 set
Product Code. 17147781
Brand: STEMCELL Technologies 17874RF
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
  • The EasySep™ HLA Chimerism Whole Blood CD19 Positive Selection Kit is designed to isolate CD19+ cells from whole blood or buffy coat by positive selection
  • Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD19 and dextran-coated magnetic particles
  • The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • The CD19 antigen is present on mature B cells and B cell progenitors but is lost upon maturation to plasma cells
  • It is also expressed on follicular dendritic cells
  • This product replaces the EasySep™ Human Whole Blood CD19 Positive Selection Kit (Catalog #18084) for even faster cell isolations
Specifications
Cell Type B Cells
Quantity 60 mL Buffy Coat or Whole Blood