missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies RoboSep™ Human B Cell Isolation Kit

Product Code. 17167821
Encompass
Change view
Click to view available options
Quantity:
1 x 109 cells
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17167821 1 x 109 cells 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17167821 Supplier STEMCELL Technologies Supplier No. 17954RF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips

  • The EasySep™ Human B Cell Isolation Kit is designed to isolate B cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection
  • The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles
  • The magnetically labeled cells are separated from the untouched, desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
  • This product can be used in place of the EasySep™ Human B Cell Enrichment Kit (Catalog #19054) for even faster cell isolations

Specifications

Cell Type B Cells
For Use With (Application) Cell Isolation
Quantity 1 x 109 cells
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.