missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies RoboSep™ Release Human CD45 Positive Selection Kit for Humanized Mice
Click to view available options
Quantity:
1 x 109 Cells
Unit Size:
1 set
Description
- The EasySep™ Release Human CD45 Positive Selection Kit is designed to isolate CD45+ leukocytes from single-cell suspensions of primary human tissues and tumors, or tissues and tumor xenografts from humanized mice, by immunomagnetic positive selection
- Desired cells are labeled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet
- Unwanted cells are simply poured off, while desired cells remain in the tube
- Then, bound magnetic particles are removed from the EasySep™-isolated CD45+ cells, which are immediately available for downstream applications
- Following cell isolation with this EasySep™ Release kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins or other chemically related ligands
Specifications
Specifications
| Cell Type | Leukocytes |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 Cells |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction