Learn More
SP Genevac™ Rocket Synergy 2 ASE Tube Puck Assembly
Product Code. 31723025
Change view
Click to view available options
Quantity:
6/Set
Unit Size:
Pack of 6
Product Code. Quantity Unit Size
31723025 6/Set Pack of 6
1 options
Product Code. 31723025 Supplier SP Genevac™ Supplier No. 702000
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Set of 6 ASE Tube Puck Assembly's
  • Allows up to 18 Dionex™ tubes to be processed simultaneously
  • Set of 6
  • Compatible with Genevac Rocket Synergy
Specifications
Type Tube Puck Assembly
Quantity 6/Set