Learn More
SP Genevac™ Rocket Synergy 2 ASE Tube Puck
Product Code. 18847477
Change view
Click to view available options
Type:
Adapter Set
Product Code. Type
18847477 Adapter Set
1 options
Product Code. 18847477 Supplier SP Genevac™ Supplier No. 702036
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
ASE Tube Puck with Flip-Flop Assembly with perfluoroelastomer seals.
  • Set of 18
  • For use with ASE tube puck (Genevac P/N:70-2000)
Specifications
Type Adapter Set