Learn More
SP Genevac™ Rocket Synergy 2 GC Vial System
Product Code. 18837477
Change view
Click to view available options
Material:
Plastic
Product Code. Material
18837477 Plastic
1 options
Product Code. 18837477 Supplier SP Genevac™ Supplier No. 701424
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Set of 6 GC Vial Sytem Flask Assemblies
  • Uses perfluoroelastomer Seals
  • Set of 6
  • Compatible with Genevac Rocket Synergy
Specifications
Material Plastic
Type Beaker Flask