Learn More
SP Genevac™ Rocket Synergy 2 Outer Lid Seal
Product Code. 31723017
Change view
Click to view available options
Material:
EPDM
Unit Size:
Each
Product Code. Material Unit Size
31723017 EPDM Each
1 options
Product Code. 31723017 Supplier SP Genevac™ Supplier No. 045818/S
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Replacement seal for the for the outer Lid of the Genevac Rocket Synergy 2 Benchtop Evaporator
  • Replacement Seal
  • Compatible with Rocket Synergy 2 Rocket Synergy Rocket 4D Synergy and Pre Synergy Rocket Evaporators
Specifications
Material EPDM
Type Seal