Learn More
SP Genevac™ Rocket Synergy 2 Standard Flask Assemby
Product Code. 18827477
Change view
Click to view available options
Material:
Plastic
Product Code. Material
18827477 Plastic
1 options
Product Code. 18827477 Supplier SP Genevac™ Supplier No. 701321
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Set of 6 standard flask holders with collar pad and collar.
  • Set of 6
  • Provided with Collar Pad and Collar
  • Used with Flask Rotor
  • Compatible with Genevac Rocket Synergy
Specifications
Material Plastic
Type Beaker Flask