missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rohamere 6661-0 (Mixture of Regiomers), TRC
Specifications
Specifications
| Chemical Name or Material | 2-((3,5,5-trimethyl-6-(2-(2-methylprop-2-enoyloxy)ethoxycarbonylamino)hexyl)carbamoyloxy)ethyl 2-methylprop-2-enoate |
| CAS | 1342259-25-9 |
| Molecular Formula | C23 H38 N2 O8 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C23H38N2O8/c1-16(2)19(26)30-10-12-32-21(28)24-9-8-18(5)14-23(6,7)15-25-22(29)33-13-11-31-20(27)17(3)4/h18H,1,3,8-15H2,2,4-7H3,(H,24,28)(H,25,29) |
| SMILES | CC(CCNC(=O)OCCOC(=O)C(=C)C)CC(C)(C)CNC(=O)OCCOC(=O)C(=C)C |
| IUPAC Name | 4,13-dihydroxy-7,9,9-trimethyl-3,14-dioxa-5,12-diazahexadeca-4,12-diene-1,16-diyl bis(2-methylacrylate) 4,13-dihydroxy-7,7,9-trimethyl-3,14-dioxa-5,12-diazahexadeca-4,12-diene-1,16-diyl bis(2-methylacrylate) |
| Molecular Weight (g/mol) | 470.556 |
| Formula Weight | 470.263 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?