missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Rollout Drawers

Product Code. 16227741 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Additional SS Drawer
Complete SS Drawer Set
Capacity (English):
11.5 cu. ft.
23.3 cu. ft.
29.2 cu. ft.
51.1 cu. ft.
78.8 cu. ft.
No. of Drawers:
1
6
7
12
Unit Size:
1 unit
Each
Pack of 12
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Capacity (English) No. of Drawers Unit Size
16227741 Complete SS Drawer Set 51.1 cu. ft. 12 Pack of 12
13493918 Complete SS Drawer Set 23.3 cu. ft. 6 Pack of 6
15383985 Additional SS Drawer 78.8 cu. ft. 1 Each
17277256 Complete SS Drawer Set 29.2 cu. ft. 6 Each
17798044 Additional SS Drawer 29.2 cu. ft. 1 Each
13203219 Complete SS Drawer Set 11.5 cu. ft. 7 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16227741 Supplier Thermo Scientific™ Supplier No. 6700
  • ONLINE EXCLUSIVE PRICE
    5195.26 €
    4440.93 € / Pack of 12
    Save 754.33 €
    14% Off
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Choose stainless-steel Thermo Scientific™ Rollout Drawers as a convenient alternative to open-wire refrigerator shelving.

Choose stainless-steel Thermo Scientific™ Rollout Drawers as a convenient alternative to open-wire shelving. This is an option for Thermo Scientific™ high-performance refrigerators.

  • Stainless-steel drawers
  • Adjustable with full extension

Ordering Information:

Factory installed; must be specified when ordering the refrigerator

TRUSTED_SUSTAINABILITY

Specifications

Type Complete SS Drawer Set
Capacity (English) 51.1 cu. ft.
Capacity (Metric) 1447 L
Drawers Stainless Steel
No. of Drawers 12
For Use With (Equipment) 51 cu. ft. (1447L) Refrigerators and Freezers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.