missing translation for 'onlineSavingsMsg'
Learn More

Romidepsin, TRC

Product Code. 30418988
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30418988 10 mg -
30426531 1 mg 1mg
30427956 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30418988 Supplier Toronto Research Chemicals Supplier No. TRCR42506010MG
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128517-07-7
Molecular Formula C24 H36 N4 O6 S2
Molecular Weight (g/mol) 540.7
Synonym Cyclo[(2Z)-2-amino-2-butenoyl-L-valyl-(3S,4E)-3-hydroxy-7-mercapto-4-heptenoyl-D-valyl-D-cysteinyl], cyclic (3→5)-disulfide, L-Valine, N-(3-hydroxy-7-mercapto-1-oxo-4-heptenyl)-D-valyl-D-cysteinyl-(Z)-2,3-didehydro-2-aminobutanoyl-, (4→1)-lactone, cyclic (1→2)-disulfide, [S-(E)]-, 2-Oxa-12,13-dithia-5,8,20,23-tetraazabicyclo[8.7.6]tricosane, cyclic peptide deriv., Antibiotic FR 901228, Chromadax, FK 228, FR 901228, Istodax, NSC 630176, Romidepsin
IUPAC Name (1S,4S,7Z,10S,16E,21R)-7-ethylidene-4,21-di(propan-2-yl)-2-oxa-12,13-dithia-5,8,20,23-tetrazabicyclo[8.7.6]tricos-16-ene-3,6,9,19,22-pentone
SMILES C\C=C\1/NC(=O)[C@H]2CSSCC\C=C\[C@H](CC(=O)N[C@H](C(C)C)C(=O)N2)OC(=O)[C@@H](NC1=O)C(C)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C24H36N4O6S2/c1-6-16-21(30)28-20(14(4)5)24(33)34-15-9-7-8-10-35-36-12-17(22(31)25-16)26-23(32)19(13(2)3)27-18(29)11-15/h6-7,9,13-15,17,19-20H,8,10-12H2,1-5H3,(H,25,31)(H,26,32)(H,27,29)(H,28,30)/b9-7+,16-6-/t15-,17-,19-,20+/m1/s1
Formula Weight 540.2076
Chemical Name or Material Romidepsin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.