missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ropinirole-d14 HCl (di-n-propyl-d14), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Ropinirole-D14 (N,N-dipropyl-D14) Hydrochloride |
| CAS | 1276197-10-4 |
| Molecular Formula | C16 D14 H10 N2 O . Cl H |
| Quantity | 0.005 g |
| Synonym | 4-[2-[Di(propyl-1,1,2,2,3,3,3-d7)amino]ethyl]-1,3-dihydro-2 H-indol-2-one hydrochloride, Ropinirole-d14 hydrochloride, Ropinirole-D14 (N,N-dipropyl-D14) hydrochloride, 4-[2-[Bis(1,1,2,2,3,3,3-Heptadeuteriopropyl)amino]ethyl]-1,3-dihydro-2 H-indol-2-one hydrochloride |
| InChI Formula | InChI=1 S/C16H24N2O.ClH/c1-3-9-18(10-4-2)11-8-13-6-5-7-15-14(13)12-16(19)17-15;/h5-7 H,3-4,8-12H2,1-2H3,(H,17,19);1 H/i1D3,2D3,3D2,4D2,9D2,10D2; |
| SMILES | Cl.[2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])N(CCc1cccc2NC(=O)Cc12)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 4-[2-[bis(1,1,2,2,3,3,3-heptadeuteriopropyl)amino]ethyl]-1,3-dihydroindol-2-one;hydrochloride |
| Molecular Weight (g/mol) | 310.922 |
| Formula Weight | 310.253 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction