missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ropivacaine hydrochloride, MedChemExpress
Description
Ropivacaine hydrochloride is a potent sodium channel blocker and blocks impulse conduction via reversible inhibition of sodium ion influx in nerve fibrese. Ropivacaine is also an inhibitor of K2P (two-pore domain potassium channel) TREK-1 with an IC50 of 402.7 μM in COS-7 cell's membrane. Ropivacaine is widely used for neuropathic pain management in vivo.
Specifications
Specifications
| Chemical Name or Material | Ropivacaine hydrochloride |
| CAS | 98717-15-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C17H27ClN2O |
| Quantity | 50 mg |
| Solubility Information | DMSO : 10 mg/mL (32.17 mM; Need ultrasonic) ∣H2O : 10 mg/mL (32.17 mM; Need ultrasonic) |
| SMILES | [H]Cl.O=C([C@H]1N(CCC)CCCC1)NC2=C(C)C=CC=C2C |
| Molecular Weight (g/mol) | 310.86 |
| Formula Weight | 310.86 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?