Learn More
ROR gamma/RORC/NR1F3 Mouse anti-Human, Mouse, Clone: 600214, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 2 publications
Brand:  R&D Systems MAB6109-SP
Encompass
Product Code. 15727824
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
ROR gamma/RORC/NR1F3 Monoclonal specifically detects ROR gamma/RORC/NR1F3 in Human, Mouse samples. It is validated for Western Blot.