missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROR gamma t/RORC2/NR1F3 Rabbit anti-Human, Clone-1181A, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB61093-100
This item is not returnable.
View return policy
Description
ROR gamma t/RORC2/NR1F3 Monoclonal specifically detects ROR gamma t/RORC2/NR1F3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| ROR gamma t/RORC2/NR1F3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 6097 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Flow Cytometry | |
| 1181A | |
| Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells | |
| P51449 | |
| E.coli derived recombinant human ROR gamma T/RORC2/NR1F3 Met1-Gln100 Accession # P51449 | |
| 100 μg | |
| Primary | |
| Detects recombinant human ROR gamma t/RORC2/NR1F3 by direct ELISAs. | |
| Human | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction