missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rose Bengal, 85%, pure, high purity, Biological stain

Product Code. 13478428 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
1g
25g
500g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13478428 100 g Glass bottle 100g
10571441 1 g Glass bottle 1g
10730891 5 g Glass bottle 5g
10504654 25 g Glass bottle 25g
13408438 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 13478428 Supplier Thermo Scientific Chemicals Supplier No. 189451000
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 632-69-9 | C20H4Cl4I4Na2O5 | 1019.644 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-69-9
Molecular Formula C20H4Cl4I4Na2O5
Molecular Weight (g/mol) 1019.644
MDL Number MFCD00005043
InChI Key DPBQSRZQYAYDGY-UHFFFAOYSA-N
Synonym Acid Red 94, C.I. 45440
PubChem CID 87244310
IUPAC Name sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na]

Specifications

Absorption Ratio 1.21 to 1.95
Dye Content 80% min.
Infrared Spectrum Authentic
Lambda Max 540 to 550nm (in water)
Loss on Drying 10% max. (105°C)
Packaging Glass bottle
Assay Percent Range 85%
Color Bordeaux/Red to Brown/Red
Solubility Information Solubility in water: soluble (36%).
Quantity 100 g
Flash Point >92°C
Physical Form Crystalline Powder
Formula Weight 1017.65
Percent Purity 85%
Grade Pure
Chemical Name or Material Rose Bengal
Show More Show Less
compliance-icons
Product Identifier
  • Rose Bengal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.