missing translation for 'onlineSavingsMsg'
Learn More

Rose Bengal, Pure, C.I.45440, Fisher Chemical

Product Code. 10172490 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10172490 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10172490 Supplier Fisher Chemical Supplier No. R/0400/46

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 632-69-9
Molecular Formula C20H4Cl4I4Na2O5
MDL Number 151169
InChI Key DPBQSRZQYAYDGY-UHFFFAOYSA-N
SMILES C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na]
IUPAC Name sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one
Molecular Weight (g/mol) 1019.644
PubChem CID 87244310
Formula Weight 1017.65g/mol
Packaging Glass Jar
Color Brown/Red
pH 7 to 10
Quantity 25 g
Physical Form Powder
Show More Show Less
Product Identifier
  • Rose Bengal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.