missing translation for 'onlineSavingsMsg'
Learn More

Roseoflavin, ≥97%, MP Biomedicals™

Product Code. 11494591 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
0.1 g
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494591 0.1 g 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11494591 Supplier MP Biomedicals Supplier No. 0215425280
 Restricted Item
Estimated Shipment: 05-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Antibacterial

Chemical Identifiers

CAS 51093-55-1
Molecular Formula C18H23N5O6
Molecular Weight (g/mol) 405.411
InChI Key IGQLDUYTWDABFK-GUTXKFCHSA-N
Synonym roseoflavin, roseoflavine, 8-demethyl-8-dimethylamino riboflavin, 1-deoxy-1-8-dimethylamino-7-methyl-2,4-dioxo-3,4-dihydrobenzo g pteridin-10 2h-yl-d-ribitol
PubChem CID 49867612
ChEBI CHEBI:72346
IUPAC Name 8-(dimethylamino)-7-methyl-10-[(2S,3S,4R)-2,3,4,5-tetrahydroxypentyl]benzo[g]pteridine-2,4-dione
SMILES CC1=CC2=C(C=C1N(C)C)N(C3=NC(=O)NC(=O)C3=N2)CC(C(C(CO)O)O)O

Specifications

Color Brown
Assay Percent Range ≥97%
Quantity 0.1 g
Formula Weight 405.4
Physical Form Solid
Chemical Name or Material Roseoflavin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.