missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies RosetteSep™ HLA Granulocyte Depletion Cocktail

Product Code. 17117711
Encompass
Change view
Click to view available options
Quantity:
1000 mL whole blood
250 mL whole blood
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17117711 1000 mL whole blood 1 set
17197701 250 mL whole blood 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17117711 Supplier STEMCELL Technologies Supplier No. 15684HLA

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunodensity depletion of granulocytes for HLA analysis

  • The RosetteSep™ HLA Granulocyte Depletion Cocktail is designed to deplete granulocytes from whole blood.
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD66b and glycophorin A on red blood cells (RBCs).
  • When centrifuged over a buoyant density medium such as Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs.
  • The desired cells are present as a highly enriched population depleted of granulocytes at the interface between the plasma and the buoyant density medium.

Note: In light of the new EU Regulation 2017/746 (IVDR) of the European Parliament and of the Council on in vitro diagnostic medical devices, which is expected to become effective in 2022, STEMCELL recently deregistered and removed the CE IVD claims associated with these products, and they are now labeled ;For Research Use Only. Please note there have been no changes to the form, function and quality assurances associated with these products.

Specifications

Cell Type Granulocytes and Subsets
For Use With (Application) Cell Isolation; In Vitro Diagnostic
Quantity 1000 mL whole blood
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.