missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Rosiglitazone
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
- Rosiglitazone is a potent and selective PPARγ ligand
- It binds to the PPARγ ligand-binding domain with a Kd of 43 nM (Lehmann et al.) It activates luciferase-based expression constructs PPARγ1 and PPARγ2 with EC50 values of approximately 30 nM and 100 nM, respectively (Lehmann et al.)
- Stimulates embryonic mouse neural stem cell proliferation and inhibits neuronal differentiation (Wada et al.)
- Induces adipocyte differentiation in C3H10T1/2 stem cells (Lehmann et al.)
- Promotes endothelial differentiation, and inhibits smooth muscle differentiation, in angiogenic progenitor cells (Wang et al.)
- Stimulates adipocyte differentiation, and decreases osteogenesis, in mouse and human bone marrow-derived mesenchymal stem cells (Ali et al.; Benvenuti et al.; Lecka-Czernik et al.; Sorocéanu et al.)
- Enhances osteoclast formation in mouse bone marrow cells stimulated with RANKL and M-CSF (Wu et al.)
Specifications
Specifications
| CAS | 122320-73-4 |
| Synonym | Avandia; BRL 49653 |
| Quantity | 10 mg |
| Molecular Formula | C18H19N3O3S |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction