Learn More
Tocris Bioscience™ Rosiglitazone
GREENER_CHOICE
Potent and selective PPARγ agonist; antidiabetic agent
Brand:  Tocris Bioscience™ 5325/10
Encompass
Product Code. 15786675
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Rosiglitazone is a potent and selective PPARγ agonist (EC50 = 60 nM); exhibits no activity at PPARα and PPARβ. Promotes differentiation of pluripotent C3H10T1/2 stem cells into adipocytes. Also promotes differentiation of urothelial organoids in combination with erlotinib. Exhibits antihyperglycemic activity in diabetic ob/ob mouse model. Antidiabetic agent.
Specifications
Specifications
5-[[4-[2-(Methyl-2-pyridinylamino)ethoxy]phenyl]methyl]-2,4-thiazolidinedione
122320-73-4
BRL 49653