missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Rotarus™ smart 30 set with PKF 60-16-3

Product Code. 16457039
Change view
Click to view available options
Channels:
1
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Channels unitSize
16457039 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16457039 Supplier Hirschmann™ Supplier No. 950113260

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High quality peristaltic pump set consisting of a PKF 60-series single-channel pump head and an exact smart 30 feed pump, which can be used in combination with regular stopper-less tubes.

  • Precise smart 30 delivery pump
  • Ideal for low-viscosity media
  • Speed can be programmed directly on the panel and TFT touchscreen on the housing front
  • Simple and easy loading of the tubing is given by unlocking the pump head
  • Digital interface
  • For regular stopper-less tubes with a WT of 1.6 mm
  • The pump head is made of durable PPS, a high-performance plastic with very high heat resistance, high chemical resistance and durability.

Specifications

Accuracy Approx. 5%
Certifications/Compliance Machinery Directives 2006/42/EC, EMC Directives 2014/30 EC
Channels 1
Color White, Black
Control Type TFT touchscreen, no external control module
Flow Rate 2.2 to 1500 mL/min. per channel
Reversible Yes
External Control Digital Input
External Control-Input Start/Stop, rotation direction
For Use With Tubes with 1.6 mm wall thickness and inner diameter of 1.6 to 8.0 mm
Material Pump drive: steel white powder coated (IP43); Pump head: Housing PPS
Motor Type DC Motor
No. of Rollers 3
Dimensions (L x W x H) Pump drive: 190 x 83 x 220 mm; Pump head: 54 x 94 x 106 mm
Overall Length (Metric) Pump drive: 190 mm; Pump head: 54 mm
Overall Width (Metric) Pump drive: 83 mm; Pump head: 94 mm
Overall Height (Metric) Pump drive: 220 mm; Pump head: 106 mm
Voltage 100/240 VAC
Wattage 30 W
Amperage 100 VA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.