Learn More
Fisherbrand™ Rotary Mixer Discs
The range of accessories can also be autoclaved and allow for easy loading and tube removal.
Brand:  Fisherbrand™
Code : 03
Additional Details : Weight : 0.01000kg
Product Code. 12398448
  • 121.00 € / Each
Estimated Shipment: 19-06-2024
to see stock.
Add to basket
Description
Description
  • Rotary Mixer Disc plain ready to accept Flask 'Pick n Mix' or Tube 'Pick n Mix' clips
  • Can be autoclaved and allow for easy loading and tube removal
  • The accessory is easy to fit onto the RM-1 locating on three pins then secured in place with thumb wheel
Specifications
Specifications
Rotary Mixer (RM-1 and RM-2)