Learn More
Testo™ Rotating Vane Anemometer
Brand: Testo™ 0560 4170
Code : A0
Additional Details : Weight : 0.70000kg
Specifications
Vane Anemometer | |
277 x 105 x 45 mm | |
-20 to +70°C |
±0.5 m/sec. | |
Measurement of volumetric flow, flow velocity and temperature |