missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rotigotine

Product Code. 17291242 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 17291242

Brand: Thermo Scientific Chemicals 466545000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99755-59-6 | C19H25NOS, C19H25NOS | 315.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99755-59-6
Molecular Formula C19H25NOS

Specifications

Quantity 500 mg
Formula Weight 315.47
InChI Key KFQYTPMOWPVWEJ-UHFFFAOYNA-N
SMILES CCCN(CCC1=CC=CS1)C1CCC2=C(O)C=CC=C2C1
IUPAC Name 6-{propyl[2-(thiophen-2-yl)ethyl]amino}-5,6,7,8-tetrahydronaphthalen-1-ol
Molecular Weight (g/mol) 315.48
Chemical Name or Material Rotigotine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.