Learn More
Eppendorf Rotor for Centrifuge, 5425/5425 R
Product Code. 31908730 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
24 x 1.5/2.0 mL Tubes
Unit Size:
Each
Product Code. Capacity Unit Size
31908730 24 x 1.5/2.0 mL Tubes Each
1 options
Product Code. 31908730 Supplier Eppendorf Supplier No. 5495518002
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Rotor for centrifuge including lid.
  • Max. speed: 21,300 x g (15,060 rpm)
  • Max. capacity: 24 x 1.5/2.0 mL tubes
  • Aerosol-tight with Eppendorf QuickLock™ Pro lid
TRUSTED_SUSTAINABILITY
Specifications
Material Aluminum
Type Fixed Angle Rotor with Aerosol-tight Lid
Capacity 24 x 1.5/2.0 mL Tubes
Max. RCF 21,300 x g
Max. Speed 15,060 rpm
For Use With (Equipment) Centrifuge 5425/5425R
Certifications/Compliance IVD
Includes Rotor lid with QuickLock Pro
No. of Places 24
Biocontainment Aerosol-tight
Quantity 1