Learn More
Eppendorf™ Rotor Lids
Product Code. 10791755 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
A-2-MTP
Centrifuge 5427R, FA-45-24-11-Kit
Centrifuge 5427R, FA-45-30-11
F-34-6-38
F-45-17-17-Cyro
F-45-30-11
F-45-48-11
F-45-64-5-PCR
FA-45-16-17
FA-45-24-11
FA-45-24-11-HS
FA-45-24-11-Kit
FA-45-30-11
FA-45-48-11
S-24-11-AT
Material:
Aluminum
Aluminum, Coated
Polypropylene
Description:
Aerosol-tight
Aerosol-tight, QuickLock
Coated
Non-aerosol-tight
Wind shield upper shell
Unit Size:
Each
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Material Description unitSize
10791755 F-34-6-38 Aluminum Non-aerosol-tight Each
12334948 A-2-MTP Aluminum Wind shield upper shell Each
15234536 Centrifuge 5427R, FA-45-24-11-Kit Aluminum Aerosol-tight Each
15244536 Centrifuge 5427R, FA-45-30-11 Aluminum Aerosol-tight Each
10197523 F-45-17-17-Cyro Polypropylene Non-aerosol-tight Each
15254536 F-45-30-11 Polypropylene Non-aerosol-tight Each
10005693 F-45-30-11 Polypropylene Coated Each
10782135 F-45-64-5-PCR Aluminum Non-aerosol-tight Each
15224536 FA-45-24-11 Aluminum Aerosol-tight Each
13226589 FA-45-16-17 Aluminum Aerosol-tight Each
10279323 FA-45-24-11-HS Aluminum Aerosol-tight Each
13246938 F-45-48-11 Polypropylene Non-aerosol-tight Each
10269753 FA-45-24-11-Kit Aluminum Aerosol-tight Each
13286948 FA-45-24-11-Kit Aluminum Aerosol-tight, QuickLock Each
10072383 FA-45-30-11 Aluminum Aerosol-tight Each
10496132 FA-45-30-11 Aluminum Non-aerosol-tight Each
13216958 FA-45-30-11 Aluminum Aerosol-tight, QuickLock Each
10076213 FA-45-30-11 Aluminum, Coated Aerosol-tight Each
13246958 FA-45-48-11 Aluminum Aerosol-tight, QuickLock Each
15274536 FA-45-48-11 Aluminum Aerosol-tight Each
15284536 FA-45-48-11 Polypropylene Non-aerosol-tight Each
13236948 S-24-11-AT Aluminum Aerosol-tight Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
Product Code. 10791755 Supplier Eppendorf™ Supplier No. 5804727509
  • ONLINE EXCLUSIVE PRICE
    202.00 €
    198.00 € / Pack of 1
    Save 4.00 €
    1% Off
In Stock
Add to basket
Add to basket
Designed for use with Eppendorf Rotors
  • Replacement rotor lids
  • Aerosol-tight and aluminum lids available
  • Available in a variety of sizes
TRUSTED_SUSTAINABILITY
Specifications
Material Aluminum
For Use With (Equipment) F-34-6-38
Description Non-aerosol-tight
Product Type Rotor Lid